Learn More
Enzo Life Sciences HSP60 (insect) polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA805F
Encompass_Preferred
Host: Rabbit. Immunogen: Native Hsp60 from Heliothis virescens (insect) sperm. UniProt ID: P25420. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Sheep, Scallop, Rabbit, Porcine, Mussel, Moth, Monkey, Lobster, Hamster, Guinea pig, Grasshopper, Fish, Ehrlichia, E. coli, Drosophila, Dog, Crab, Coral, Cockroach, Chicken, Bovine, Beluga. Applications: IF, WB. Recommended Dilutions: Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7483