Learn More
Enzo Life Sciences HSP60 monoclonal antibody (LK-1) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPA806D
Encompass_Preferred
Clone: LK-1. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human Hsp60. Source: Purified from ascites. Species reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Drosophila, Dog, Chicken, Bovine. Applications: ELISA, Flow Cytometry, IF, IHC (PS), IP, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7484