Learn More
Enzo Life Sciences HSP60 monoclonal antibody (Mab11-13) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA829F
Encompass_Preferred
Clone: Mab-11-13. Host: Mouse. Isotype: IgG2a. Immunogen: Recombinant human Hsp60. Source: Purified from ascites. Species reactivity: Human, Mouse, Rat Snake, Rabbit, Porcine, Mosquito, Monkey, Hamster, Guinea pig, Fish, Drosophila, Dolphin, Dog, Bovine. Applications: Flow Cytometry, IF, IHC (PS), WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7533