Learn More
Enzo Life Sciences HSP60 monoclonal antibody (LK-2) (100 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA807E
Encompass_Preferred
Clone: LK-2. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human HSP60. UniProt: P10809. Source: Purified from ascites. Species Reactivity: Human, Mouse, Rat, Ehrlichia sennetsu, Rabbit, Plant, Porcine, Monkey, Hamster, Guinea pig, Dog, Chicken, C. elegans, Bovine. Applications: ELISA. Flow Cytometry, IHC, IP, WB. Recommended dilutions: Flow Cytometry (10µg/ml). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.1mM PMSF and 50% glycerol.
Catalog No. 50-667-544