Learn More
Medchemexpress LLC HSP60 Mouse N-His 10ug
SDP
Catalog No. 5000179267
Encompass_Preferred
The HSP60 protein together with Hsp10 contributes to the import and assembly of mitochondrial proteins It ensures correct folding and prevents incorrect folding under stressful conditions HSP60 Protein Mouse (N-His) is the recombinant mouse-derived HSP60 expressed by E coli with N-10 His labeled tag The total length of HSP60 Protein Mouse (N-His) is 572 a a
Catalog No. 50-001-79267 Supplier Medchemexpress LLC Supplier No. HYP73916A10UG
The HSP60 protein together with Hsp10 contributes to the import and assembly of mitochondrial proteins It ensures correct folding and prevents incorrect folding under stressful conditions HSP60 Protein Mouse (N-His) is the recombinant mouse-derived HSP60 expressed by E coli with N-10 His labeled tag The total length of HSP60 Protein Mouse (N-His) is 572 a a