Learn More
Enzo Life Sciences HSP60 (low endotoxin) (mouse), (recombinant) (200µg)
SDP
Supplier:  Enzo Life Sciences ADIESP741F
Encompass_Preferred
Alternative name: Chaperonin 60, CPN60, HspD1, Heat shock protein 60. Endotoxin: <50EU/mg purified protein (LAL test). Purity: ≥90% (SDS-PAGE; Western blot). MW: ~60kDa. Formulation: Liquid. In 10mM TRIS, pH 7.5, containing 150mM sodium chloride, 1mM EDTA, and 1mM DTT. Source: Produced in E. coli. Long Term Storage: -80°C. Applications: ELISA, WB Activity assay.
Catalog No. 50-200-7228