Learn More
Enzo Life Sciences HSP60 polyclonal antibody (1mg)
SDP
Supplier:  Enzo Life Sciences ADISPA828J
Encompass_Preferred
Host: Goat. Immunogen: Recombinant human Hsp60. UniProt ID: P10809. Source: Purified from goat serum. Species reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Porcine, Monkey, Mycobacterium bovis, Hamster, Guinea pig, Fish, Drosophila, Dog, Chicken, Bovine, Beluga. Applications: IHC, IP, WB. Recommended Dilutions: Immunoprecipitation (1:100) Western Blot (1:1,000, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7531