Learn More
Enzo Life Sciences HSP60 polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPA828D
Encompass_Preferred
Host: Goat. Immunogen: Recombinant human Hsp60. Source: Purified from goat serum. Species reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Porcine, Monkey, Mycobacterium bovis, Hamster, Guinea pig, Fish, Drosophila, Dog, Chicken, Bovine, Beluga. Applications: IHC, IP, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7529