Learn More
Enzo Life Sciences HSP 60 (human), (recombinant) (100 µg)
SDP
Supplier:  Enzo Life Sciences ADINSP540E
Encompass_Preferred
Alternative name: Chaperonin 60. CPN60. HspD1. Heat shock protein 60. MW: ~60kDa. Endotoxin content: >500EU/mg purified protein (LAL test). Purity: ≥90% (SDS-PAGE; Western blot). Formulation: Liquid. In Dulbecco's PBS. Source: Produced in E. coli. UniProt: P10809. Application: ELISA, WB, in vitro Assay, Antigen microarray, Activity assay. Recommended dilutions: Western Blot (100ng, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Long Term Storage: -80°C
Catalog No. 50-666-992