Learn More
Enzo Life Sciences HSP 60 (mouse), (recombinant) (50 µg)
SDP
Supplier:  Enzo Life Sciences ADISPP741D
Encompass_Preferred
Alternative name: Chaperonin 60, CPN60, HspD1, Heat shock protein 60. MW: ~60kDa. Purity: ≥90% (SDS-PAGE; Western blot). Formulation: Liquid. In 10mM TRIS-HCl, pH 7.5, containing 150mM sodium chloride, 1.0mM EDTA, and 1.0mM DTT. Source: Produced in E. coli. UniProt: P63038. Application: ELISA, WB, Activity assay. Long Term Storage: -80°C
Catalog No. 50-667-751