Learn More
Enzo Life Sciences HSP60 (rat), (recombinant) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPP742F
Encompass_Preferred
Alternative name: Chaperonin 60, CPN60, HspD1, Heat shock protein 60. Purity: ≥85% (SDS-PAGE; Western blot). MW: ~60kDa. Formulation: Liquid. In 50mM TRIS-HCl, pH 7.5, containing 150mM sodium chloride, 0.1mM PMSF, and 1.0mM DTT. Source: Produced in E. coli. Long Term Storage: -80°C. Applications: WB in vitro Assay, Activity assay.
Catalog No. 50-200-7629