Learn More
Cayman Chemical Hsp60HspD1 PolycLNal ntI 100ug
SDP
Catalog No. 502935219
Encompass_Preferred
An antineoplastic agent inhibits DNA replication and cell proliferation and induces cell cycle arrest in the G1/S phase inhibits proliferation of NCI H460 human lung cancer cells in vitro (IC50 0.56 mM) reduces RNR activity by 75 when used at a concentration of 5 mM via degradation of its active site tyrosyl radical reduces leukocyte adhesion and extravasation in a humanized mouse model of SCD via NO production and a cGMP-dependent pathway inhibits HIV-1 viral replication in PBMCs (IC90 0.4 mM)
Catalog No. 50-293-5219 Supplier Cayman Chemical Supplier No. 24531100 UG
An antineoplastic agent inhibits DNA replication and cell proliferation and induces cell cycle arrest in the G1/S phase inhibits proliferation of NCI H460 human lung cancer cells in vitro (IC50 0.56 mM) reduces RNR activity by 75 when used at a concentration of 5 mM via degradation of its active site tyrosyl radical reduces leukocyte adhesion and extravasation in a humanized mouse model of SCD via NO production and a cGMP-dependent pathway inhibits HIV-1 viral replication in PBMCs (IC90 0.4 mM)