Learn More
Enzo Life Sciences HSP65 (low endotoxin) (M. bovis), (recombinant) (50µg)
SDP
Supplier:  Enzo Life Sciences ADIESP581D
Encompass_Preferred
Alternative name: Heat shock protein 65. Endotoxin: <50EU/mg purified protein (LAL test). Purity: ≥95% (SDS-PAGE). MW: ~65kDa. Formulation: Liquid. In 1x Dulbecco's PBS. Source: Produced in E. coli. Long Term Storage: -80°C. Applications: WB Antigen microarray, Activity assay.
Catalog No. 50-200-7224