Learn More
Enzo Life Sciences HSP65 (mycobacterial) monoclonal antibody (3F7) (200µl)
SDP
Supplier:  Enzo Life Sciences ADISPA881F
Encompass_Preferred
Clone: 3F7. Host: Mouse. Isotype: IgM. Immunogen: Recombinant M. bovis HSP65. UniProt ID: P0A521. Source: Purified from mouse ascites. Species reactivity: Human, Mouse, Rat Mycobacterium leprae, Mycobacterium bovis, Hamster, Guinea pig, E. coli, Drosophila, Cryptosporidium, Bovine. Applications: WB. Recommended Dilutions: Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Ammonium sulfate precipitation purified. Formulation: Liquid. In PBS containing 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7571