Learn More
Enzo Life Sciences HSP65 (mycobacterial) monoclonal antibody (4H11) (100µg)
SDP
Supplier:  Enzo Life Sciences ADISPA882E
Encompass_Preferred
Clone: 4H11. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant M. bovis BCG Hsp65. UniProt ID: P0A521. Source: Purified from mouse ascites. Species reactivity: Mycobacterium leprae, Mycobacterium bovis, Bovine. Applications: WB in vitro Assay. Recommended Dilutions: Western Blot (1µg/ml, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7572