Learn More
PHYTO AB INC HSP70 Antibody Immunogen: AT3G12580 Background: Hsp70s is molecular chaperones and play a crucial role in plant response to stress conditions, and in plant growth and development. 150ug/EA
Catalog No. NC2877316
Encompass_Preferred
PhytoAB Inc. is a contract research organization and plant antibody R&D based services and products supplier. PhytoAB is privately owned company headquatered in the San Francisco Bay Area of California, USA. PhytoAB specializes in antibody production for plant scientic usage and customized services, focus on the current need of plant antibodies to assist the further investigation of the current booming results from deep genome sequencing and increasing protomics studies. We have specialized R&D and customer services teams to fulfill the needs of botany & plant sciences research and industry world wide with high quality, professional and cutting edge services, and offering complete products usage guidance and suggestions for related problem solutions.
Catalog No. NC2877316 Supplier PHYTO AB INC Supplier No. PHY0167
PhytoAB Inc. is a contract research organization and plant antibody R&D based services and products supplier. PhytoAB is privately owned company headquatered in the San Francisco Bay Area of California, USA. PhytoAB specializes in antibody production for plant scientic usage and customized services, focus on the current need of plant antibodies to assist the further investigation of the current booming results from deep genome sequencing and increasing protomics studies. We have specialized R&D and customer services teams to fulfill the needs of botany & plant sciences research and industry world wide with high quality, professional and cutting edge services, and offering complete products usage guidance and suggestions for related problem solutions.