Learn More
Enzo Life Sciences HSP70-A1 (low endotoxin) (mouse), (recombinant) (200µg)
SDP
Supplier:  Enzo Life Sciences ADIESP502F
Encompass_Preferred
Alternative name: Heat shock protein 70. Endotoxin: <50EU/mg purified protein (LAL test). Purity: ≥90% (SDS-PAGE; Western blot). MW: ~70kDa (observed). Formulation: Liquid. In 1x Dulbecco’s PBS, pH 7.4, containing 5.0mM glutathione. Source: Produced in E. coli. Long Term Storage: -80°C. Applications: WB Activity assay.
Catalog No. 50-200-7220