Learn More
Enzo Life Sciences HSP70 ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADIEKS700B
Encompass_Preferred
Widely used ELISA HSP70 kit for chaperone and stress research. The HSP70 EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 4.5 hours. Absorbance is read at 450 nm. This kit is ideal for measuring intracellular HSP70 (HSP72). Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis. Alternative Name: Heat shock protein 70. HSP72. Sensitivity: 200 pg/ml (range 780 - 50,000 pg/ml). Assay Time: 4.5 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Human, Mouse, Rat. Contents: Microtiter plate, 5X Extraction reagent, Standard, Sample diluent 2, Wash buffer concentrate, Antibody, Conjugate, TMB Substrate, Stop solution 2
Catalog No. 50-659-133