Learn More
Enzo Life Sciences HSP70 (fish) polyclonal antibody (200µl)
SDP
Supplier:  Enzo Life Sciences ADISPA763F
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant salmon Hsp70. UniProt ID: Q91233. Source: Whole rabbit serum. Species reactivity: Human, Mouse, Rat Fish, Drosophila, Dog, Chicken, Bovine. Applications: WB. Recommended Dilutions: Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Antiserum. Formulation: Liquid. Whole rabbit serum. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7463