Learn More
Enzo Life Sciences HSP 70 HSP 72 monoclonal antibody (C92F3A-5) (200 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA810F
Encompass_Preferred
Clone: C92F3A-5. Host: Mouse. Isotype: IgG1. Immunogen: Native human Hsp70 protein. UniProt: P0DMV8 (HSPA1A), P0DMV9 (HSPA1B). Source: Purified from ascites. Species Reactivity: Human, Mouse, Rat, Xenopus, Teal, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Gerbil, Fish, Drosophila, Dog, Chicken, C. elegans, Bovine, Beluga. Applications: ELISA. Flow Cytometry, ICC, IF, IHC, IP, WB, Electron microscopy. Recommended dilutions: Immunohistochemistry (1:50). Western Blot (1:1,000). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-554