Learn More
Enzo Life Sciences HSP70/HSP72 monoclonal antibody (C92F3A-5) (PE conjugate) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA810PEF
Encompass_Preferred
Clone: C92F3A-5. Host: Mouse. Isotype: IgG1. Immunogen: Native human Hsp70. UniProt ID: P0DMV8 (HSPA1A), P0DMV9 (HSPA1B). Source: Purified from ascites. Species reactivity: Human, Mouse, Rat, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Fish, Drosophila, Dog, Chicken, Bovine, Beluga. Applications: Flow Cytometry. Recommended Dilutions: Flow Cytometry (1:100) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 0.09% sodium azide. Handling: Avoid vigorous centrifugation and vortexing. Do not freeze. Protect from light.
Catalog No. 50-200-7495