Learn More
Enzo Life Sciences HSP70/HSP72 monoclonal antibody (C92F3A-5) (AP conjugate) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPA810APD
Encompass_Preferred
Clone: C92F3A-5. Host: Mouse. Isotype: IgG1. Immunogen: Native human Hsp70. Source: Purified from ascites. Species reactivity: Human, Mouse, Rat Teal, Fish, Dog, Chicken, Bovine. Applications: ELISA, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7489