Learn More
Enzo Life Sciences HSP70/HSP72 monoclonal antibody (C92F3A-5) (FITC conjugate) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPA810FID
Encompass_Preferred
Clone: C92F3A-5. Host: Mouse. Isotype: IgG1. Immunogen: Native human Hsp70. Source: Purified from ascites. Species reactivity: Human, Mouse, Rat, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Fish, Drosophila, Dog, Chicken, Bovine. Applications: Flow Cytometry. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide. Handling: Avoid freeze/thaw cycles. Protect from light.
Catalog No. 50-200-7492