Learn More
Enzo Life Sciences HSP70/HSP72 monoclonal antibody (C92F3A-5) (1000µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6921000
Encompass_Preferred
Clone: C92F3A-5. Isotype: IgG1. Immunogen: Native human Hsp70 protein. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Mouse, Rat, Xenopus, Teal, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Gerbil, Fish, Drosophila, Dog, Chicken, C. elegans, Bovine, Beluga. Applications: ELISA, Flow Cytometry, ICC, IF, IHC, IP, WB Electron microscopy. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-201-3229