Learn More
Enzo Life Sciences HSP70/HSP72 monoclonal antibody (C96F3-3) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA8132F
Encompass_Preferred
Clone: C96F3-3. Host: Mouse. Isotype: IgG2b. Immunogen: Native human Hsp70. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Mouse, Rat Fish, Bovine. Applications: WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7501