Learn More
Enzo Life Sciences HSP70/HSP72 monoclonal antibody (N31F2-4) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPA8131D
Encompass_Preferred
Clone: N31F2-4. Host: Mouse. Isotype: IgG1. Immunogen: Native human Hsp70. UniProt ID: P0DMV8 (HSPA1A), P0DMV9 (HSPA1B). Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human E. coli. Applications: WB. Recommended Dilutions: Western Blot (1:1,000, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7498