Learn More
Enzo Life Sciences HSP70/HSP72 polyclonal antibody (1mg)
SDP
Supplier:  Enzo Life Sciences ADISPA812J
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human Hsp70. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Sheep, Porcine, Monkey, Hamster, Guinea pig, Fish, Drosophila, Dog, Bovine, Beluga. Applications: ICC, IF, IHC, IP, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7497