Learn More
Enzo Life Sciences HSP70/HSP72 polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA811F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human Hsp70. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Sheep, Porcine, Monkey, Hamster, Dog, Bovine, Beluga. Applications: IP, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7496