Learn More
Enzo Life Sciences HSP70/HSP72 (rat), (recombinant) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPP758D
Encompass_Preferred
Alternative name: Heat shock protein 70, HspA1A, HspA1B. Purity: ≥90% (SDS-PAGE; Western blot). MW: ~72kDa. Formulation: Liquid. In 50mM TRIS, pH 7.5, containing 1.0mM DTT, 100mM sodium chloride, and 0.1mM PMSF. Source: Produced in E. coli. Long Term Storage: -80°C. Applications: WB Proliferation assay, GST pulldown, Activity assay.
Catalog No. 50-200-7633