Learn More
Enzo Life Sciences HSP70/HSP72 (Chinook salmon), (recombinant) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPP763F
Encompass_Preferred
Alternative name: Heat shock protein 70, HspA1A, HspA1B. Purity: ≥90% (SDS-PAGE; Western blot). MW: ~70kDa. Formulation: Liquid. In Dulbecco's PBS. Source: Produced in E. coli. Long Term Storage: -80°C. Applications: WB Activity assay.
Catalog No. 50-200-7637