Learn More
Enzo Life Sciences HSP70 (low endotoxin) (human), (recombinant) (50µg)
SDP
Supplier:  Enzo Life Sciences ADIESP555D
Encompass_Preferred
Low endotoxin HSP70 recombinant protein. Alternative name: Heat shock protein 70, HspA1A, HspA1B, HSP72. Endotoxin: <50EU/mg purified protein (LAL test). Purity: ≥90% (SDS-PAGE; Western blot). MW: ~72kDa. Formulation: Liquid. In Dulbecco's PBS. Source: Produced in E. coli. Long Term Storage: -80°C. Applications: ELISA, WB in vitro Assay, Activity assay.
Catalog No. 50-200-7223