Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hsp70-in-1 | 1268273-90-0 | 98.1% | 464.58 g/mol | C24H28N6O2S | 10MG
SDP

Catalog No. 5000239554
Encompass_Preferred

HSP70-IN-1 is a small-molecule inhibitor of the heat shock protein HSP70 developed for research applications. It inhibits Kasumi-1 cell growth with an IC50 of 2.3 μM and is supplied as a light yellow solid or as a 10 mM solution in DMSO. The compound is used in cellular assays to probe HSP70 function, cell-cycle regulation, and proteostasis-related pathways.

  • Selective HSP70 inhibition for cellular assays.
  • IC50 2.3 μM in Kasumi-1 cell growth assays.
  • CAS number 1268273-90-0 for unambiguous identification.
  • Molecular formula C24H28N6O2S; molecular weight 464.58 g/mol.
  • High purity, 98.07% by supplier analysis.
  • Appearance solid, light yellow to yellow.
  • Soluble in DMSO, 50 mg/mL (≈107.6 mM); ultrasonic assistance recommended.
  • Available in small research quantities and as a 10 mM DMSO solution.

Catalog No. 50-002-39554 Supplier Medchemexpress LLC Supplier No. HY1262210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HSP70-IN-1 is a small-molecule inhibitor of the heat shock protein HSP70 developed for research applications. It inhibits Kasumi-1 cell growth with an IC50 of 2.3 μM and is supplied as a light yellow solid or as a 10 mM solution in DMSO. The compound is used in cellular assays to probe HSP70 function, cell-cycle regulation, and proteostasis-related pathways.

  • Selective HSP70 inhibition for cellular assays.
  • IC50 2.3 μM in Kasumi-1 cell growth assays.
  • CAS number 1268273-90-0 for unambiguous identification.
  • Molecular formula C24H28N6O2S; molecular weight 464.58 g/mol.
  • High purity, 98.07% by supplier analysis.
  • Appearance solid, light yellow to yellow.
  • Soluble in DMSO, 50 mg/mL (≈107.6 mM); ultrasonic assistance recommended.
  • Available in small research quantities and as a 10 mM DMSO solution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.