Learn More
Enzo Life Sciences HSP70B’ ELISA (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADIEKS725A
Encompass_Preferred
Only commercially available ELISA kit for ultra-sensitive HSP70B’ detection in cell lysates, serum, and tissue. The HSP70B’ EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 4.5 hours. Absorbance is read at 450 nm. Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis. Alternative name: Heat shock protein 70, HSP72. Sensitivity: 62 pg/ml (range 0.156 - 10 ng/ml). Use/Stability: Store all components at 4°C, except standard at -20°C. UniProt ID: P17066. Assay Time: <4.5 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human . Content: Microtiter plate, 5X Extraction reagent, Standard, Sample diluent, Wash buffer concentrate, Antibody, Antibody diluent, HRP Conjugate, Conjugate diluent, TMB Substrate, Stop solution 2.
Catalog No. 50-200-7217