Learn More
Enzo Life Sciences HSP70B' monoclonal antibody (165f) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA754F
Encompass_Preferred
Clone: 165f. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of human HSP70B'. Source: Purified from mouse ascites. Species reactivity: Human. Applications: WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7457