Learn More
Enzo Life Sciences HSP70B' monoclonal antibody (165f) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPA754D
Encompass_Preferred
Clone: 165f. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of human HSP70B'. UniProt ID: P17066. Source: Purified from mouse ascites. Species reactivity: Human. Applications: WB. Recommended Dilutions: Western Blot (1:1,000, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7456