Learn More
Enzo Life Sciences HSP70B' polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADISPA756E
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of human Hsp70B'. Source: Purified from rabbit serum. Species reactivity: Human, Mouse. Applications: IHC, IP, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7458