Learn More
Enzo Life Sciences HSP71 (mycobacterial) monoclonal antibody (5A8) (1mg)
SDP
Supplier:  Enzo Life Sciences ADISPA885J
Encompass_Preferred
Clone: 5A8. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant mycobacterial Hsp71. Source: Purified from mouse ascites. Species reactivity: Mycobacterium tuberculosis. Applications: WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7573