Learn More
Medchemexpress LLC Hsp90 alpha/beta Antibody (YA356) | 90 kDa | 100 UL
SDP
Catalog No. 5000395888
Encompass_Preferred
Hsp90 alpha/beta Antibody (YA356) is a rabbit-derived, non-conjugated IgG monoclonal antibody that targets Hsp90 alpha/beta. Its purity is affinity purified, and it is supplied in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol, and 0.05% BSA with 0.01% Sodium azide as a preservative. The antibody has an observed band size of 90 kDa.
  • Rabbit host, reactive with human, mouse, and rat samples.
  • Non-conjugated.
  • Stable for 1 year when stored at -20°C; avoid repeated freeze/thaw cycles.
  • Liquid appearance.
  • Shipped with blue ice.
  • Applicable in signal transduction research.
  • Validated for western blot (WB), immunohistochemistry-paraffin (IHC-P), immunohistochemistry-frozen (IHC-F), immunocytochemistry/immunofluorescence (ICC/IF), and immunoprecipitation (IP).
Catalog No. 50-003-95888 Supplier Medchemexpress LLC Supplier No. HYP80714100UL
Hsp90 alpha/beta Antibody (YA356) is a rabbit-derived, non-conjugated IgG monoclonal antibody that targets Hsp90 alpha/beta. Its purity is affinity purified, and it is supplied in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol, and 0.05% BSA with 0.01% Sodium azide as a preservative. The antibody has an observed band size of 90 kDa.
  • Rabbit host, reactive with human, mouse, and rat samples.
  • Non-conjugated.
  • Stable for 1 year when stored at -20°C; avoid repeated freeze/thaw cycles.
  • Liquid appearance.
  • Shipped with blue ice.
  • Applicable in signal transduction research.
  • Validated for western blot (WB), immunohistochemistry-paraffin (IHC-P), immunohistochemistry-frozen (IHC-F), immunocytochemistry/immunofluorescence (ICC/IF), and immunoprecipitation (IP).