Learn More
Enzo Life Sciences HSP090alpha (human), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADIEKS895
Encompass_Preferred
Most sensitive, rapid ELISA kit for highly specific, quantitative detection of human HSP90α. The HSP90α (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 450 nm. Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis. Alternative Name: Heat shock protein 90 alpha. HSP86. Sensitivity: 50 pg/ml (range 62.5 - 4,000 pg/ml). Assay Time: <3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Human. Contents: Microtiter plate, 5X Extraction reagent 2, Standard, Sample diluent, Wash buffer concentrate, HRP Conjugate, Conjugate diluent, TMB Substrate, Stop solution 2
Catalog No. 50-659-139