Learn More
Enzo Life Sciences HSP90alpha (human), (recombinant) (50 µg)
SDP
Supplier:  Enzo Life Sciences ADISPP776D
Encompass_Preferred
Alternative name: HSP86, Heat shock protein 90alpha. MW: ~90kDa. Purity: ≥90% (SDS-PAGE; Western blot). Formulation: Liquid. In Dulbecco's PBS containing 2.7mM potassium chloride, 1.5mM potassium phosphate, 137mM sodium chloride, 8.1mM sodium phosphate, and 10% glycerol. Source: Produced in E. coli. UniProt: P07900. Application: WB. Long Term Storage: -80°C
Catalog No. 50-667-775