Learn More
Enzo Life Sciences HSP 90 monoclonal antibody (16F1) (50 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA835D
Encompass_Preferred
Clone: 16F1. Host: Rat. Isotype: IgG2a. Immunogen: Native human Hsp90. UniProt: P07900 (HSP90alpha), P08238 (HSP90beta). Source: Purified from ascites. Species Reactivity: Human, Mouse, Rat, Xenopus, Sheep, Scallop, Rabbit, Plant, Porcine, Opossum, Mussel, Monkey, Hamster, Guinea pig, Fish, Drosophila, Dog, Chicken, Bovine, Beluga. Applications: IHC (PS), IP, WB. Recommended dilutions: Western Blot (1:1,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-624