Learn More
Abcam Recombinant human Hsp90 beta protein (Active), 100UG
Supplier:  Abcam AB80033100UG
Encompass_Preferred
Recombinant human Hsp90 beta protein (Active) is a Human Full Length protein, in the 1 to 724 aa range, expressed in Baculovirus infected BTI-TN-5B1-4, with >90% purity and suitable for western blot, SDS-PAGE, ELISA, Functional studies and more.The predicted molecular weight of ab80033 protein is 90 kDa. - Save time and ensure accurate results - use our Hsp90 beta protein as a control - Optimal protein bioactivity, stability and reproducibility - Available in different sizes to fit your experimental needs
Protein/ Peptide/ Enzyme Synonyms: HSP90B, HSPC2, HSPC3, HSPCB, HSP90AB1, Heat shock protein HSP 90-beta, HSP 90, Heat shock 84 kDa, Heat shock protein family C member 3, HSP 84, HSP84
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1824815