Learn More
Enzo Life Sciences HSP90 (human), (native) (200µg)
SDP
Catalog No. 502007644
Encompass_Preferred
Alternative name: HSP86, Heat shock protein 90. Purity: ≥90% (SDS-PAGE; Western blot). MW: ~90kDa. Formulation: Liquid. In 50mM TRIS-HCl, pH 7.5, containing 1.0mM DTT and 100mM sodium chloride. Source: Produced in HeLa cells. Long Term Storage: -80°C. Applications: ELISA, WB Antigen microarray.
Catalog No. 50-200-7644 Supplier Enzo Life Sciences Supplier No. ADISPP770F
Alternative name: HSP86, Heat shock protein 90. Purity: ≥90% (SDS-PAGE; Western blot). MW: ~90kDa. Formulation: Liquid. In 50mM TRIS-HCl, pH 7.5, containing 1.0mM DTT and 100mM sodium chloride. Source: Produced in HeLa cells. Long Term Storage: -80°C. Applications: ELISA, WB Antigen microarray.