Learn More
Medchemexpress LLC Hsp90-IN-17 hydrochloride | 1253584-63-2 | 99.3% | 476.87 | 50 MG
SDP
Catalog No. 5000361476
Encompass_Preferred
Hsp90-IN-17 hydrochloride is an HSP90 inhibitor used in the study of proliferative diseases, including cancer and neurodegenerative diseases. It has an IC50 of 0.362 μM against human A2780 cells in an antiproliferative activity assay.
  • Can be used in the study of proliferative diseases
  • Exhibits antiproliferative activity against human A2780 cells
  • Suitable for research use only
Catalog No. 50-003-61476 Supplier Medchemexpress LLC Supplier No. HY148215A50MG
Hsp90-IN-17 hydrochloride is an HSP90 inhibitor used in the study of proliferative diseases, including cancer and neurodegenerative diseases. It has an IC50 of 0.362 μM against human A2780 cells in an antiproliferative activity assay.
  • Can be used in the study of proliferative diseases
  • Exhibits antiproliferative activity against human A2780 cells
  • Suitable for research use only