Learn More
Medchemexpress LLC Hsp90-IN-37 10mM 1mL
SDP
Supplier:  Medchemexpress LLC HYW10793510MM/1ML
Encompass_Preferred
Hsp90-IN-37 (Z-2) is a heat shock protein 90 (Hsp90) inhibitor that inhibits Hsp90 enzymatic activity by 69% Hsp90-IN-37 has antitumor activity[1]
Catalog No. 50-002-05087