Learn More
Enzo Life Sciences HSP90 monoclonal antibody (2D11B9) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA831200
Encompass_Preferred
Clone: 2D11B9. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human Hsp90. UniProt ID: P07900 (HSP90α), P08238 (HSP90β). Source: Purified from ascites. Species reactivity: Human, Mouse, Rat, Sheep, Porcine, Monkey, Horse, Hamster, Guinea pig, Fish, Drosophila, Dog. Applications: WB. Recommended Dilutions: Western Blot (1µg/ml, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7539