Learn More
Enzo Life Sciences HSP90 monoclonal antibody (2D12) (200µl)
SDP
Supplier:  Enzo Life Sciences ADISPA845F
Encompass_Preferred
Clone: 2D12. Host: Rat. Isotype: IgM. Immunogen: Native mouse Hsp90. Source: Purified from mouse ascites. Species reactivity: Human, Mouse, Rat Water mold, Sheep, Rabbit, Monkey, Hamster, Guinea pig, Fish, Dog, Chicken, Bovine, Beluga. Applications: IP, WB. Purity: Ammonium sulfate precipitation purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7561