Learn More
Enzo Life Sciences HSP90 monoclonal antibody (3G3) (400µl)
SDP
Supplier:  Enzo Life Sciences ALX804079R400
Encompass_Preferred
Clone: 3G3. Isotype: IgM. Immunogen: Purified mouse HSP90 (heat shock protein 90). Species reactivity: Human, Mouse, Rat Rainbow trout, Chicken. Applications: IF, IHC, IP, WB. Purity: Ammonium sulfate/ascites suspension. Formulation: Liquid. Contains no preservatives.
Catalog No. 50-200-9773