Learn More
Enzo Life Sciences HSP90 monoclonal antibody (AC88) (DyLight 488 conjugate) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA830488F
Encompass_Preferred
Clone: AC88. Host: Mouse. Isotype: IgG1. Immunogen: Achlya ambisexualis (water mold) Hsp90. UniProt ID: Q8LLI6. Source: Purified from ascites. Species reactivity: Human, Mouse, Rat Fusobacterium nucleatum, Water mold, Sheep, Scallop, Rabbit, Porcine, Mussel, Monkey, Mycobacterium leprae, Hamster, Guinea pig, Fish, E. coli, Dog, Chicken, C. elegans, Beluga. Applications: Flow Cytometry. Recommended Dilutions: Flow Cytometry (1:10) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 0.09% sodium azide. Handling: Avoid freeze/thaw cycles. Protect from light.
Catalog No. 50-200-7535