Learn More
Enzo Life Sciences HSP90beta monoclonal antibody (K3705) (200 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA842F
Encompass_Preferred
Clone: K3705. Host: Mouse. Isotype: IgM. Immunogen: Recombinant human HSP90β. UniProt: P08238. Source: Purified from hybridoma tissue culture supernatant. Species Reactivity: Human, Mouse, Rat, Sheep, Porcine, Hamster, Guinea pig, Dog, Chicken, Bovine. Applications: IHC, WB. Recommended dilutions: Immunohistochemistry (1:50). Western Blot (1:1,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Ammonium sulfate precipitation purified. Formulation: Liquid. In saline, pH 6.9, containing 0.1% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-635